Learn More
Aat Bioquest EGTA AM CAS 99590-86-0 (10 mg)
SDP
Catalog No. 502249902
Encompass_Preferred
EGTA AM is cell-permeable version of EGTA ethylene glycol tetraacetic acid a cell-permeable calcium chelator. This catalog is for 10 mg.
This item is not returnable. View return policy
Catalog No. 50-224-9902 Supplier Aat Bioquest Supplier No. 21005
EGTA AM is cell-permeable version of EGTA ethylene glycol tetraacetic acid a cell-permeable calcium chelator. This catalog is for 10 mg.