Learn More
Medchemexpress LLC EGTA-AM (EGTA Acetoxymethyl ester) | 99590-86-0 | 99.8% | 1 MG
SDP
Catalog No. 5000409749
Encompass_Preferred
EGTA-AM is a membrane permeable form of EGTA, which can be passively loaded into cells to generate intracellular EGTA. It is also a Ca2+ chelator with slow chelating dynamics. For research use only.
  • Membrane permeable, allowing passive loading into cells.
  • Generates intracellular EGTA.
  • Ca2+ chelator with slow chelating dynamics.
Catalog No. 50-004-09749 Supplier Medchemexpress LLC Supplier No. HYD09731MG
EGTA-AM is a membrane permeable form of EGTA, which can be passively loaded into cells to generate intracellular EGTA. It is also a Ca2+ chelator with slow chelating dynamics. For research use only.
  • Membrane permeable, allowing passive loading into cells.
  • Generates intracellular EGTA.
  • Ca2+ chelator with slow chelating dynamics.