Learn More
Alkali Scientific EGTA [Ethylene glycol bis(2 aminoethylether) N,N,N',N' tetraacetic acid], 25 G
SDP
Catalog No. 5000659673
Encompass_Preferred
EGTA (Ethylene glycol bis(2-aminoethylether) N,N,N',N' tetraacetic acid) is a powerful calcium chelator commonly used in laboratory applications. This 25g package is ideal for researchers working with cell biology, biochemistry, and enzymology. EGTA binds calcium ions with high specificity and is often used in experiments that require the removal of calcium to study calcium-dependent processes, such as enzyme activities or cellular signaling. The high affinity of EGTA for calcium makes it a preferred reagent in studies involving the regulation of intracellular calcium levels. It is also commonly used in preparation buffers for experiments involving cellular and molecular assays.
Catalog No. 50-006-59673 Supplier Alkali Scientific Supplier No. GTA002CP
EGTA (Ethylene glycol bis(2-aminoethylether) N,N,N',N' tetraacetic acid) is a powerful calcium chelator commonly used in laboratory applications. This 25g package is ideal for researchers working with cell biology, biochemistry, and enzymology. EGTA binds calcium ions with high specificity and is often used in experiments that require the removal of calcium to study calcium-dependent processes, such as enzyme activities or cellular signaling. The high affinity of EGTA for calcium makes it a preferred reagent in studies involving the regulation of intracellular calcium levels. It is also commonly used in preparation buffers for experiments involving cellular and molecular assays.