Learn More
Alkali Scientific EGTA [Ethylene glycol bis(2 aminoethylether) N,N,N',N' tetraacetic acid], 250 Grams
SDP
Catalog No. 5000659601
Encompass_Preferred
EGTA (Ethylene glycol bis(2-aminoethylether) N,N,N',N'-tetraacetic acid) is a chelating agent that binds calcium ions, making it essential for molecular biology and biochemical research. This reagent is widely used in studies involving calcium signaling, where precise control over calcium levels is crucial. EGTA effectively binds calcium ions without interfering with magnesium ions, which is important for enzyme reactions and protein studies. This 250-gram bottle is suitable for laboratory use where high purity and calcium ion regulation are needed. It is particularly useful in studies on cellular signaling, protein activity assays, and enzyme inhibition, where accurate ion control is required.
Catalog No. 50-006-59601 Supplier Alkali Scientific Supplier No. GTA006CP
EGTA (Ethylene glycol bis(2-aminoethylether) N,N,N',N'-tetraacetic acid) is a chelating agent that binds calcium ions, making it essential for molecular biology and biochemical research. This reagent is widely used in studies involving calcium signaling, where precise control over calcium levels is crucial. EGTA effectively binds calcium ions without interfering with magnesium ions, which is important for enzyme reactions and protein studies. This 250-gram bottle is suitable for laboratory use where high purity and calcium ion regulation are needed. It is particularly useful in studies on cellular signaling, protein activity assays, and enzyme inhibition, where accurate ion control is required.