Learn More
Alkali Scientific EGTA [Ethylene glycol bis(2 aminoethylether) N,N,N',N' tetraacetic acid], 50 G
SDP
Catalog No. 5000659703
Encompass_Preferred
EGTA (Ethylene glycol bis(2-aminoethylether) N,N,N',N' tetraacetic acid) is an effective calcium chelator, with this 50g package commonly used in laboratory settings for various biochemical and cell biology applications. EGTA has a strong affinity for calcium ions and is particularly useful in studies that require calcium removal to analyze calcium-dependent processes, such as enzyme activity and cell signaling. This reagent is commonly employed in buffers for cell culture studies, molecular assays, and in the preparation of solutions for experiments involving calcium regulation. Its ability to selectively chelate calcium ions makes it indispensable for experiments requiring precise control of calcium concentrations.
Catalog No. 50-006-59703 Supplier Alkali Scientific Supplier No. GTA003CP
EGTA (Ethylene glycol bis(2-aminoethylether) N,N,N',N' tetraacetic acid) is an effective calcium chelator, with this 50g package commonly used in laboratory settings for various biochemical and cell biology applications. EGTA has a strong affinity for calcium ions and is particularly useful in studies that require calcium removal to analyze calcium-dependent processes, such as enzyme activity and cell signaling. This reagent is commonly employed in buffers for cell culture studies, molecular assays, and in the preparation of solutions for experiments involving calcium regulation. Its ability to selectively chelate calcium ions makes it indispensable for experiments requiring precise control of calcium concentrations.