Learn More
Hach Company EGTA Solution, 50 mL SCDB
Supplier:  Hach Company 2229726
Encompass
For calcium and magnesium hardness determination by the Calmagite colorimetric method. Hach method 8030. 50 mL Self-Contained Dropping Bottle
Catalog No. 50-199-5583