Learn More
AnaSpec EGTA, tetrasodium salt, 10 mM aqueous solution *UltraPure Grade*, | MW 468.3 | (10 mL)
Catalog No. 50850378
Encompass
EGTA, tetrasodium salt, 10 mM aqueous solution *UltraPure Grade*, ; MW 468.3; (10 mL) A chelating agent useful for the determination of calcium in the presence of magnesium
Catalog No. 50-850-378 Supplier AnaSpec Supplier No. AS84097
EGTA, tetrasodium salt, 10 mM aqueous solution *UltraPure Grade*, ; MW 468.3; (10 mL) A chelating agent useful for the determination of calcium in the presence of magnesium