Learn More
Labtech Inc EH35B hotplate, Heating surface: Stainless Steel, Temperature range: Ambient - 350℃, Heating power: 2000W, Temperature Accuracy: +/- 2.0 C, Power Supply: 230V, 50/60Hz
Catalog No. 5000496191
Encompass_Preferred
EH35B hotplate, Heating surface: Stainless Steel, Temperature range: Ambient - 350℃, Heating power: 2000W, Temperature Accuracy: +/- 2.0 C, Power Supply: 230V, 50/60Hz
Catalog No. 50-004-96191 Supplier Labtech Inc Supplier No. LH2050
EH35B hotplate, Heating surface: Stainless Steel, Temperature range: Ambient - 350℃, Heating power: 2000W, Temperature Accuracy: +/- 2.0 C, Power Supply: 230V, 50/60Hz