Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Eh44

Catalog No. 9063951G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
Catalog No. Quantity
90-639-51G 1 g
1 options

Catalog No. 90-639-51G

Supplier: Sigma Aldrich 9063951G

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

EH44 is a carbazole derivative used as interfacial, hole transporting and contact layer in perovskite solar cell (PSC). Using EH44 can increase the performance and the lifetime of the devices. EH44 is a Li+ free, hydrophobic hole transporting material (HTM) which can eliminate degradation due to Li+ migration, supresse degradation due to moisture absorption and has long-term stability in fabricated PSC.EH44 is a Li+ free, hydrophobic hole transporting material (HTM). It eliminates degradation due to Li+ migration, supresses degradation due to moisture absorption and thus showed promise for achieving long-term stability in fabricated perovskite solar cells. Using EH44 as HTM in a device configuration of: SnO2/PAL/EH44/MoOx /Al, not only generated high power conversion efficiency (PCE) comparable to the record PCE by using the state-of-the-art-HTM Spiro-OMeTAD, but also most importantly showed greatly improved device stability: even under the combined stresses of light (including ultraviolet light), oxygen and moisture, perovskite solar cells using EH44 as hole transporting layer were shown to retain 94% of their peak efficiency despite 1,000 hours of continuous unencapsulated operation in ambient air conditions (relative humidity of 10-20%). When stability tests were conducted in an inert atmosphere, degradation of only GO+2% over the course of 1,500 hours of continuous operation was observed.Note that in the above mentioned result, silver bis(trifluoromethanesulfonyl)imide (product 668001) was used to oxidize EH44 and yield EH44+TFSI.EH44+TFSI was then blended with neat EH44 at various ratios to control the conductivity (and optical transparency if desired) and optimize the device performance. The impressive power conversion efficiency (PCE) of 18.5% was achieved for a perovskite solar cell using EH44 layers of GO+60 nm thickness doped with 14wt% EH44+TFSI and 25 mg/ml 4-tert-butylpyridine (product 142379), which is comparable to device performance using spiro-OMeTAD (19%) as the HTM.

Specifications

CAS 1879076-81-9
Molecular Formula C48H51N3O4
Linear Formula C48H51N3O4
Quantity 1 g
Synonym 9-(2-Ethylhexyl)-N,N,N,N-tetrakis(4-methoxyphenyl)- 9 H-carbazole-2,7-diamine)
Molecular Weight (g/mol) 733.94
Percent Purity ≥99% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.