Learn More
Labtech Inc EH45APlus hotplate, Heating surface: Graphite, Temperature range: Ambient - 450℃, Heating power: 2800W, Temperature Accuracy: +/- 5.0 C, Power Supply: 230V, 50/60Hz
Catalog No. 5000496192
Encompass_Preferred
EH45APlus hotplate, Heating surface: Graphite, Temperature range: Ambient - 450℃, Heating power: 2800W, Temperature Accuracy: +/- 5.0 C, Power Supply: 230V, 50/60Hz
Catalog No. 50-004-96192 Supplier Labtech Inc Supplier No. LH2101
EH45APlus hotplate, Heating surface: Graphite, Temperature range: Ambient - 450℃, Heating power: 2800W, Temperature Accuracy: +/- 5.0 C, Power Supply: 230V, 50/60Hz