Learn More
Labtech Inc EH45B hotplate, Heating surface: Graphite, Temperature range: Ambient - 450℃, Heating power: 2800W, Temperature Accuracy: +/- 2.0 C, Power Supply: 230V, 50/60Hz
Catalog No. 5000496177
Encompass_Preferred
EH45B hotplate, Heating surface: Graphite, Temperature range: Ambient - 450℃, Heating power: 2800W, Temperature Accuracy: +/- 2.0 C, Power Supply: 230V, 50/60Hz
Catalog No. 50-004-96177 Supplier Labtech Inc Supplier No. LH2110
EH45B hotplate, Heating surface: Graphite, Temperature range: Ambient - 450℃, Heating power: 2800W, Temperature Accuracy: +/- 2.0 C, Power Supply: 230V, 50/60Hz