Learn More
Labtech Inc EH45C hotplate, Heating surface: Graphite with Ceramic Coating, Temperature range: Ambient - 450℃, Heating power: 2800W, Temperature Accuracy: +/- 2.0 C, Power Supply: 230V, 50/60Hz
Catalog No. 5000496197
Encompass_Preferred
EH45C hotplate, Heating surface: Graphite with Ceramic Coating, Temperature range: Ambient - 450℃, Heating power: 2800W, Temperature Accuracy: +/- 2.0 C, Power Supply: 230V, 50/60Hz
Catalog No. 50-004-96197 Supplier Labtech Inc Supplier No. LH2120
EH45C hotplate, Heating surface: Graphite with Ceramic Coating, Temperature range: Ambient - 450℃, Heating power: 2800W, Temperature Accuracy: +/- 2.0 C, Power Supply: 230V, 50/60Hz