Learn More
INTACT GENOMICS INC Thymidine Auxotrophic EHA101 Agrobacterium Chemically Competent Cells 15x50µL
Catalog No. 5000082951
Encompass
Intact Genomics (ig) Thymidine-Auxotrophic EHA101Thy Agrobacterium Chemically Competent Cells include modifications so that they will not grow unless 50 mg/L of thymidine is added to Minimal medium. This prevents the bacteria from overgrowing plant tissues when used for plant transformation. Thymidine-Auxotrophic EHA101Thy Chemically Competent Agrobacterium cells are optimized for the highest transformation efficiencies. The EHA101Thy strain is useful for transgenic operations of rice, tobacco and other plants. EHA101Thy– thyA knockout mutant, spectinomycin-resistant, derived from EHA101, itself a derivative of A281 (A136/pTiBo542).Benefits: Thymidine Auxotrophic Reduced bacterial overgrowth during co-cultivation Decreased need for antibioticsProduct Components Chemically Competent Agrobacterium Cells DNA (pTF102, 500 pg/µl) Recovery medium* * Client needs to add 50 mg/L of thymidine (not provided) to recovery media before use
Catalog No. 50-000-82951 Supplier INTACT GENOMICS INC Supplier No. 130215
Intact Genomics (ig) Thymidine-Auxotrophic EHA101Thy Agrobacterium Chemically Competent Cells include modifications so that they will not grow unless 50 mg/L of thymidine is added to Minimal medium. This prevents the bacteria from overgrowing plant tissues when used for plant transformation. Thymidine-Auxotrophic EHA101Thy Chemically Competent Agrobacterium cells are optimized for the highest transformation efficiencies. The EHA101Thy strain is useful for transgenic operations of rice, tobacco and other plants. EHA101Thy– thyA knockout mutant, spectinomycin-resistant, derived from EHA101, itself a derivative of A281 (A136/pTiBo542).Benefits: Thymidine Auxotrophic Reduced bacterial overgrowth during co-cultivation Decreased need for antibioticsProduct Components Chemically Competent Agrobacterium Cells DNA (pTF102, 500 pg/µl) Recovery medium* * Client needs to add 50 mg/L of thymidine (not provided) to recovery media before use