Learn More
INTACT GENOMICS INC EHA105 Agrobacterium Chemically Competent Cells 6x50µl
Catalog No. 5000083050
Encompass
Intact Genomics EHA105 Agrobacterium Chemically Competent Cells are optimized for the highest transformation efficiencies which is ideal for applications requiring high transformation efficiencies, such as with cDNA or gDNA library construction. The EHA105 strain is useful for transgenic operations of rice, tobacco and other plants. EHA105 contains a rifampicin resistance gene (rif). EHA105 strain also contains an amber basic Ti plasmid pEHA105 (pTiBo542DT-DNA) without self-transport function, which contains the vir gene.Product Includes and Storage:EHA105 Agrobacterium Chemically Competent Cells: -80 ºCpCAMBIA1391z control DNA: -20 ºCAgro Recovery medium: 4 ºC
Catalog No. 50-000-83050 Supplier INTACT GENOMICS INC Supplier No. 108406
Intact Genomics EHA105 Agrobacterium Chemically Competent Cells are optimized for the highest transformation efficiencies which is ideal for applications requiring high transformation efficiencies, such as with cDNA or gDNA library construction. The EHA105 strain is useful for transgenic operations of rice, tobacco and other plants. EHA105 contains a rifampicin resistance gene (rif). EHA105 strain also contains an amber basic Ti plasmid pEHA105 (pTiBo542DT-DNA) without self-transport function, which contains the vir gene.Product Includes and Storage:EHA105 Agrobacterium Chemically Competent Cells: -80 ºCpCAMBIA1391z control DNA: -20 ºCAgro Recovery medium: 4 ºC