Learn More
Gold Biotechnology Inc EHA105 Agro Electro Cells
SDP
Catalog No. 5000562328
Encompass_Preferred
GoldBio s EHA105 Electrocompetent Agrobacterium cells allow you to obtain high transformation efficiency in applications such as gDNA or cDNA library construction Our EHA105 strain contains a rifampicin resistance gene (rif) and an amber basic Ti plasmid pEHA105 (pTiBo542DT-DNA) without self-transport function containing the vir gene EHA105 is ideal for genetic transformation of rice tobacco and several other plants
Catalog No. 50-005-62328 Supplier Gold Biotechnology Inc Supplier No. CC22510X50
GoldBio s EHA105 Electrocompetent Agrobacterium cells allow you to obtain high transformation efficiency in applications such as gDNA or cDNA library construction Our EHA105 strain contains a rifampicin resistance gene (rif) and an amber basic Ti plasmid pEHA105 (pTiBo542DT-DNA) without self-transport function containing the vir gene EHA105 is ideal for genetic transformation of rice tobacco and several other plants