Learn More
INTACT GENOMICS INC Methionine Auxotrophic EHA105 Agrobacterium Chemically Competent Cells 15x50µl
Catalog No. 5000082832
Encompass
Intact Genomics (ig) Methionine-Auxotrophic EHA105Met Agrobacterium Chemically Competent Cells include modifications so that they will not grow unless methionine is added to Minimal medium. This prevents the bacteria from overgrowing plant tissues when used for plant transformation. Methionine- Auxotrophic EHA105Met Agrobacterium Chemically Competent Cells are optimized for the highest transformation efficiencies. The EHA105Met strain is useful for transgenic operations of rice, tobacco and other plants. EHA105 contains a rifampicin resistance gene (rif). EHA105Met strain also contains an amber basic Ti plasmid pEHA105 (pTiBo542DT-DNA) without self-transport function, which contains the vir gene.Benefits: Methionine Auxotrophic Enables development of more efficient transformation systems Reduced bacterial overgrowth during co-cultivation Decreased need for antibiotics
Catalog No. 50-000-82832 Supplier INTACT GENOMICS INC Supplier No. 107815
Intact Genomics (ig) Methionine-Auxotrophic EHA105Met Agrobacterium Chemically Competent Cells include modifications so that they will not grow unless methionine is added to Minimal medium. This prevents the bacteria from overgrowing plant tissues when used for plant transformation. Methionine- Auxotrophic EHA105Met Agrobacterium Chemically Competent Cells are optimized for the highest transformation efficiencies. The EHA105Met strain is useful for transgenic operations of rice, tobacco and other plants. EHA105 contains a rifampicin resistance gene (rif). EHA105Met strain also contains an amber basic Ti plasmid pEHA105 (pTiBo542DT-DNA) without self-transport function, which contains the vir gene.Benefits: Methionine Auxotrophic Enables development of more efficient transformation systems Reduced bacterial overgrowth during co-cultivation Decreased need for antibiotics