Learn More
Gold Biotechnology Inc EHA105 Agrobacterium ElectroCompetent Cells 5x50 µl
SDP
Supplier:  Gold Biotechnology Inc CC2255X50C
Encompass_Preferred
GoldBio’s EHA105 ElectroCompetent Agrobacterium cells allow you to obtain high transformation efficiency in applications such as gDNA or cDNA library construction. Our EHA105 strain contains a rifampicin resistance gene (rif) and an amber basic Ti plasmid pEHA105 (pTiBo542DT-DNA) without self-transport function, containing the vir gene. EHA105 is ideal for genetic transformation of rice, tobacco and several other plants.
Catalog No. NC2385798