Learn More
Gold Biotechnology Inc EHA105 pSoup Agro Comp Cells
SDP
Catalog No. 5000562298
Encompass_Preferred
GoldBio s EHA105 (pSoup) chemically competent Agrobacterium cells are optimized for high transformation efficiencies making them ideal for applications such as cDNA or gDNA library construction These cells are resistant to rifampicin chloramphenicol and streptomycin providing versatility in selection and antibiotic resistance EHA105 strain contains a rifampicin resistance gene (rif) and an amber basic Ti plasmid pEHA105 (pTiBo542DT-DNA) without self-transport function containing the vir gene
Catalog No. 50-005-62298 Supplier Gold Biotechnology Inc Supplier No. CC11810X50
GoldBio s EHA105 (pSoup) chemically competent Agrobacterium cells are optimized for high transformation efficiencies making them ideal for applications such as cDNA or gDNA library construction These cells are resistant to rifampicin chloramphenicol and streptomycin providing versatility in selection and antibiotic resistance EHA105 strain contains a rifampicin resistance gene (rif) and an amber basic Ti plasmid pEHA105 (pTiBo542DT-DNA) without self-transport function containing the vir gene