Learn More
Gold Biotechnology Inc EHA105 pSoup Agro Electro
SDP
Catalog No. 5000562219
Encompass_Preferred
EHA105 (pSoup) Electrocompetent Agrobacterium cells are designed to provide high transformation efficiencies for applications such as cDNA or gDNA library construction The strain contains a rifampicin resistance gene (rif) and an amber basic Ti plasmid pEHA105 (pTiBo542DT-DNA) which carries the vir gene necessary for the insertion of T-DNA into the plant genome Although the pEHA105 plasmid is incapable of transferring its own T-DNA it can still transfer foreign binary vector T-DNA
Catalog No. 50-005-62219 Supplier Gold Biotechnology Inc Supplier No. CC23510X50
EHA105 (pSoup) Electrocompetent Agrobacterium cells are designed to provide high transformation efficiencies for applications such as cDNA or gDNA library construction The strain contains a rifampicin resistance gene (rif) and an amber basic Ti plasmid pEHA105 (pTiBo542DT-DNA) which carries the vir gene necessary for the insertion of T-DNA into the plant genome Although the pEHA105 plasmid is incapable of transferring its own T-DNA it can still transfer foreign binary vector T-DNA