Learn More
Gold Biotechnology Inc EHA105 pSoup P19 Agro Comp
SDP
Catalog No. 5000562259
Encompass_Preferred
GoldBio s EHA105 (pSoup-P19) chemically competent Agrobacterium cells are optimized for high transformation efficiencies making them ideal for applications such as cDNA or gDNA library construction These cells are resistant to rifampicin chloramphenicol and streptomycin providing versatility in selection and antibiotic resistance EHA105 strain contains a rifampicin resistance gene (rif) and an amber basic Ti plasmid pEHA105 (pTiBo542DT-DNA) without self-transport function containing the vir gene
Catalog No. 50-005-62259 Supplier Gold Biotechnology Inc Supplier No. CC12810X50
GoldBio s EHA105 (pSoup-P19) chemically competent Agrobacterium cells are optimized for high transformation efficiencies making them ideal for applications such as cDNA or gDNA library construction These cells are resistant to rifampicin chloramphenicol and streptomycin providing versatility in selection and antibiotic resistance EHA105 strain contains a rifampicin resistance gene (rif) and an amber basic Ti plasmid pEHA105 (pTiBo542DT-DNA) without self-transport function containing the vir gene