Learn More
Gold Biotechnology Inc EHA105 pSoup P19 Agro Electro
SDP
Catalog No. 5000562331
Encompass_Preferred
EHA105 (pSoup-P19) Electrocompetent Agrobacterium cells are designed to provide high transformation efficiencies for applications such as cDNA or gDNA library construction The strain contains a rifampicin resistance gene (rif) and an amber basic Ti plasmid pEHA105 (pTiBo542DT-DNA) which carries the vir gene necessary for the insertion of T-DNA into the plant genome Although the pEHA105 plasmid is incapable of transferring its own T-DNA it can still transfer foreign binary vector T-DNA
Catalog No. 50-005-62331 Supplier Gold Biotechnology Inc Supplier No. CC24510X50
EHA105 (pSoup-P19) Electrocompetent Agrobacterium cells are designed to provide high transformation efficiencies for applications such as cDNA or gDNA library construction The strain contains a rifampicin resistance gene (rif) and an amber basic Ti plasmid pEHA105 (pTiBo542DT-DNA) which carries the vir gene necessary for the insertion of T-DNA into the plant genome Although the pEHA105 plasmid is incapable of transferring its own T-DNA it can still transfer foreign binary vector T-DNA