Learn More
INTACT GENOMICS INC EHA105 (pSoup) Agrobacterium ElectroCompetent Cells 18x50µl
Catalog No. 5000082867
Encompass
Intact Genomics (ig) EHA105 (pSoup) Agrobacterium ElectroCompetent Cells are optimized for the highest transformation efficiencies ideal for applications such as cDNA or gDNA library construction. EHA105 (pSoup) contains a rifampicin resistance gene (rif). EHA105 (pSoup) strain also contains an amber basic Ti plasmid pEHA105 (pTiBo542DT-DNA), which contains the vir gene. The pSoup plasmid aids in replication of pGreen, 62SK, and pGs2 series plasmids. This system is used for Agrobacterium-mediated transformation of dicots such as Arabidopsis thaliana, tobacco, potatoes, and monocots like corn.Product Includes and Storage:EHA105 (pSoup) Agrobacterium ElectroCompetent Cells: -80 ºCpCAMBIA1391z control DNA: -20 ºCRecovery medium: 4 ºC
Catalog No. 50-000-82867 Supplier INTACT GENOMICS INC Supplier No. 1284PS36
Intact Genomics (ig) EHA105 (pSoup) Agrobacterium ElectroCompetent Cells are optimized for the highest transformation efficiencies ideal for applications such as cDNA or gDNA library construction. EHA105 (pSoup) contains a rifampicin resistance gene (rif). EHA105 (pSoup) strain also contains an amber basic Ti plasmid pEHA105 (pTiBo542DT-DNA), which contains the vir gene. The pSoup plasmid aids in replication of pGreen, 62SK, and pGs2 series plasmids. This system is used for Agrobacterium-mediated transformation of dicots such as Arabidopsis thaliana, tobacco, potatoes, and monocots like corn.Product Includes and Storage:EHA105 (pSoup) Agrobacterium ElectroCompetent Cells: -80 ºCpCAMBIA1391z control DNA: -20 ºCRecovery medium: 4 ºC