Learn More
Aviva Systems Biology Corp EHD1 Antibody - middle region (ARP52254_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52254_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EHD1. It was validated on Western Blot. EHD1,EH-domain containing 1,PAST; PAST1; H-PAST; HPAST1; FLJ42622; FLJ44618; EHD1,EH domain-containing protein 1,EH domain-containing protein 1,testilin,PAST homolog 1,OTTHUMP00000069747,OTTHUMP00000069748,OTTHUMP00000196087The immunogen is a synthetic peptide directed towards the middle region of human EHD1. Alternate names: FLJ42622, FLJ44618, H-PAST, HPAST1, PAST, PAST1ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-674