Learn More
Labtech Inc EHD20 Digiblock Digester, 20 positions for 100ml digestion tubes, Max Temperature: 415℃, Temp accuracy: ±1.0℃, Heating power: 2800W, Heating material: graphite coating/Al, Power Supply: 230V, 50/60Hz
Catalog No. 5000496193
Encompass_Preferred
EHD20 Digiblock Digester, 20 positions for 100ml digestion tubes, Max Temperature: 415℃, Temp accuracy: ±1.0℃, Heating power: 2800W, Heating material: graphite coating/Al, Power Supply: 230V, 50/60Hz
Catalog No. 50-004-96193 Supplier Labtech Inc Supplier No. LH7017
EHD20 Digiblock Digester, 20 positions for 100ml digestion tubes, Max Temperature: 415℃, Temp accuracy: ±1.0℃, Heating power: 2800W, Heating material: graphite coating/Al, Power Supply: 230V, 50/60Hz