Learn More
Labtech Inc EHD36S Digiblock Digester, 36 positions for 50ml digestion tubes, Max Temperature: 450℃, Temp accuracy: ±1.0℃, Heating power: 3000W, Heating material: graphite coating/Al, Power Supply: 230V, 50/60Hz
Catalog No. 5000496181
Encompass_Preferred
EHD36S Digiblock Digester, 36 positions for 50ml digestion tubes, Max Temperature: 450℃, Temp accuracy: ±1.0℃, Heating power: 3000W, Heating material: graphite coating/Al, Power Supply: 230V, 50/60Hz
Catalog No. 50-004-96181 Supplier Labtech Inc Supplier No. LH7014
EHD36S Digiblock Digester, 36 positions for 50ml digestion tubes, Max Temperature: 450℃, Temp accuracy: ±1.0℃, Heating power: 3000W, Heating material: graphite coating/Al, Power Supply: 230V, 50/60Hz