Learn More
Aviva Systems Biology Corp EHD4 Antibody - middle region (ARP55432_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55432_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EHD4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human EHD4. Alternate names: PAST4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61505