Learn More
Aviva Systems Biology Corp Ehf Antibody - middle region (ARP36843_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36843_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ehf. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 9030625L19Rik, AU019492ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-284-522