Learn More
Medchemexpress LLC HY-111904 5mg Medchemexpress, EHMT2-IN-2 CAS:2230850-47-0 Purity:>98%
SDP
Catalog No. 501932404
Encompass_Preferred
Medchemexpress, HY-111904 5mg EHMT2-IN-2 CAS:2230850-47-0 EHMT2-IN-2 is a potent EHMT inhibitor, with IC 50 s of all <100 nM for EHMT1 peptide, EHMT2 peptide and cellular EHMT2. Used in the research of blood disease or cancer [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2404 Supplier Medchemexpress LLC Supplier No. HY1119045MG
Medchemexpress, HY-111904 5mg EHMT2-IN-2 CAS:2230850-47-0 EHMT2-IN-2 is a potent EHMT inhibitor, with IC 50 s of all <100 nM for EHMT1 peptide, EHMT2 peptide and cellular EHMT2. Used in the research of blood disease or cancer [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.