Learn More
Enzo Life Sciences EHNA (50mg). CAS: 51350-19-7
SDP
Supplier:  Enzo Life Sciences BMLPD1290050
Encompass_Preferred
Phosphodiesterase PDE II inhibitor, IC50=0.8 µM. Also inhibits adenosine deaminase, Ki=4 nM. Alternative name: Erythro-9-(2-hydroxy-3-nonyl)adenine . HCl. Purity: ≥98% (HPLC). Solubility: Soluble in water (25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1377