Learn More
Medchemexpress LLC HY-103160A 2mg Medchemexpress, EHNA (hydrochloride) CAS:58337-38-5 Purity:>98%
SDP
Catalog No. 501968046
Encompass_Preferred
Medchemexpress, HY-103160A 2mg EHNA (HCl) CAS:58337-38-5 EHNA hydrochloride is a specific inhibitor of adenosine deaminase, prevents dAdo degradation and increases mitochondrial dATP levels in fibroblasts. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8046 Supplier Medchemexpress LLC Supplier No. HY103160A2MG
Medchemexpress, HY-103160A 2mg EHNA (HCl) CAS:58337-38-5 EHNA hydrochloride is a specific inhibitor of adenosine deaminase, prevents dAdo degradation and increases mitochondrial dATP levels in fibroblasts. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.