Learn More
Cayman Chemical EHNA (hydrochloride), Purity: ≥95%, CAS: 58337-38-5, 10 mg
SDP
Catalog No. 502953512
Encompass_Preferred
A dual inhibitor of adenosine deaminase (IC50 = 1.2 μM in human red blood cells) and PDE2 (IC50s = 0.8 and 2 μM from human and pig myocardium, respectively, 3.5 μM in rat hepatocyte, and 5.5 μM in human platelet)
Catalog No. 50-295-3512 Supplier Cayman Chemical Supplier No. 1335210 MG
A dual inhibitor of adenosine deaminase (IC50 = 1.2 μM in human red blood cells) and PDE2 (IC50s = 0.8 and 2 μM from human and pig myocardium, respectively, 3.5 μM in rat hepatocyte, and 5.5 μM in human platelet)