Learn More
Selleck Chemical LLC EHop-016 10mg 1380432-32-5
SDP
Supplier:  Selleck Chemical LLC S731910MG
Encompass_Preferred
EHop-016 is a specific Rac GTPase inhibitor with IC50 of 1.1 ?M for Rac1 in MDA-MB-435 and MDA-MB-231 cells, equally potent inhibition for Rac3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0619687