Learn More
TARGETMOL CHEMICALS INC EHOP-016 25MG
SDP
Catalog No. 502577124
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. EHop-016 is a specific Rac GTPase inhibitor with IC50 of 1.1 uM for Rac1 in MDA-MB-231 and MDA-MB-435 cells equally effective inhibition for Rac3. purity: 99%
Catalog No. 50-257-7124 Supplier TARGETMOL CHEMICALS INC Supplier No. T242725MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. EHop-016 is a specific Rac GTPase inhibitor with IC50 of 1.1 uM for Rac1 in MDA-MB-231 and MDA-MB-435 cells equally effective inhibition for Rac3. purity: 99%