Learn More
Abcam EHop-016, Rac GTPase inhibitor, 5MG
Catalog No. 502672668
Encompass_Preferred
MW 430.5 Da, Purity >98%. Specific Rac GTPase inhibitor. Analog of NSC-23766 (ab142161). Approximately 100-fold more potent against Rac (IC₅₀ = 1.1 μM) than NSC-23766. Specific for Rac1 and Rac3 at concentrations of ≤5 μM. At higher concentrations (10μM) inhibits the the related Rho GTPase Cdc42. Also inhibits the activity of the Rac downstream effector p21-activated kinase (PAK), lamellipodia extension, and cell migration in metastatic cancer cells.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2668 Supplier Abcam Supplier No. AB2291715MG
MW 430.5 Da, Purity >98%. Specific Rac GTPase inhibitor. Analog of NSC-23766 (ab142161). Approximately 100-fold more potent against Rac (IC₅₀ = 1.1 μM) than NSC-23766. Specific for Rac1 and Rac3 at concentrations of ≤5 μM. At higher concentrations (10μM) inhibits the the related Rho GTPase Cdc42. Also inhibits the activity of the Rac downstream effector p21-activated kinase (PAK), lamellipodia extension, and cell migration in metastatic cancer cells.
The product is subject to the following: Abcam Restricted Use Statement