Learn More
Medchemexpress LLC HY-128872 5mg Medchemexpress, EHP-101 CAS:1818428-24-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1288725MG
Encompass_Preferred
Medchemexpress, HY-128872 5mg EHP-101 CAS:1818428-24-8 EHP-101 (VCE-004.8) is an orally active, specific PPARγ and CB2 receptor dual agonist. EHP-101 inhibits prolyl-hydroxylases (PHDs) and activates the HIF pathway. EHP-101, a semi-synthetic multitarget cannabinoquinoid, has potent anti-inflammatory activity. EHP-101 attenuates adipogenesis and prevents diet-induced obesity[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0141
 Restricted Item