Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ehrlich Aldehyde Reagent, Alcoholic, for Indole Test, Ricca Chemical
SDP

Catalog No. 27904 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2790-4 120 mL
2790-16 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2790-4 Supplier Ricca Chemical Company Supplier No. 27904
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Indole Test

Specifications

Chemical Name or Material Ehrlich Aldehyde Reagent
Color Yellow
CAS 64-17-5, 7647-01-0, 7732-18-5, 100-10-7, 7732-18-5
CAS Min % 70.0, 22.96, 3.89, 0.89
CAS Max % 73.0, 23.89, 4.05, 0.93
Grade Laboratory
Packaging Amber Glass Bottle
Solvent or Matrix Alcoholic Solution
Quantity 120 mL
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.