Learn More
Medchemexpress LLC EHT 1610 | 1425945-60-3 | 98.9% | 383.40 | 100 MG
SDP
Catalog No. 5000344028
Encompass_Preferred
EHT 1610 is a potent inhibitor of DYRK family kinases, demonstrating strong activity against DYRK1A and DYRK1B. This compound exhibits antileukemia effects, modulates the cell cycle, and is capable of inducing apoptosis in various cell types. It is suitable for research applications exploring kinase inhibition and its cellular impacts.
  • Potent inhibitor of DYRK1A (IC50: 0.36 nM) and DYRK1B (IC50: 0.59 nM).
  • Exhibits antileukemia activity.
  • Regulates cell cycle progression.
  • Induces cell apoptosis.
  • Inhibits DYRK1A-mediated FOXO1 and STAT3 signaling.
  • Reduces leukemic burden in in-vivo models.
Catalog No. 50-003-44028 Supplier Medchemexpress LLC Supplier No. HY111380100MG
EHT 1610 is a potent inhibitor of DYRK family kinases, demonstrating strong activity against DYRK1A and DYRK1B. This compound exhibits antileukemia effects, modulates the cell cycle, and is capable of inducing apoptosis in various cell types. It is suitable for research applications exploring kinase inhibition and its cellular impacts.
  • Potent inhibitor of DYRK1A (IC50: 0.36 nM) and DYRK1B (IC50: 0.59 nM).
  • Exhibits antileukemia activity.
  • Regulates cell cycle progression.
  • Induces cell apoptosis.
  • Inhibits DYRK1A-mediated FOXO1 and STAT3 signaling.
  • Reduces leukemic burden in in-vivo models.