Learn More
Apexbio Technology LLC EHT 1864(Synonyms: EHT-1864, EHT1864, RAC inhibitor EHT1864, Rac GTPase inhibitor EHT 1864), 10mM (in 1mL DMSO), CAS: 754240-09-0.
SDP
Catalog No. 5000061003
Encompass_Preferred
EHT 1864 (CAS 754240-09-0) is a small-molecule inhibitor targeting Rac family GTPases with strongest affinity and inhibitory activity toward Rac1 It interacts directly with Rac isoforms Rac1 Rac1b Rac2 and Rac3 exhibiting Kd values of approximately 40 nM 50 nM 60 nM and 250 nM respectively By disrupting nucleotide binding EHT 1864 blocks Rac1 activation and prevents the formation of active Rac1/PAK-RBD complexes thus inhibiting downstream cellular functions such as PDGF-stimulated lamellipodia formation and Rac1-mediated cellular transformation This compound serves as a valuable tool in cell biology research for studying Rac-dependent signaling pathways and cellular dynamics
Catalog No. 50-000-61003 Supplier Apexbio Technology LLC Supplier No. B54875.1
EHT 1864 (CAS 754240-09-0) is a small-molecule inhibitor targeting Rac family GTPases with strongest affinity and inhibitory activity toward Rac1 It interacts directly with Rac isoforms Rac1 Rac1b Rac2 and Rac3 exhibiting Kd values of approximately 40 nM 50 nM 60 nM and 250 nM respectively By disrupting nucleotide binding EHT 1864 blocks Rac1 activation and prevents the formation of active Rac1/PAK-RBD complexes thus inhibiting downstream cellular functions such as PDGF-stimulated lamellipodia formation and Rac1-mediated cellular transformation This compound serves as a valuable tool in cell biology research for studying Rac-dependent signaling pathways and cellular dynamics