Learn More
Selleck Chemical LLC EHT 1864 2HCl
SDP
Catalog No. 5000001409
Encompass_Preferred
EHT 1864 2HCl is a potent Rac family GTPase inhibitor with Kd of 40 nM 50 nM 60 nM and 250 nM for Rac1 Rac1b Rac2 and Rac3 respectively
Catalog No. 50-000-01409 Supplier Selleck Chemical LLC Supplier No. S748250MG
Add to Cart
Add to Cart
EHT 1864 2HCl is a potent Rac family GTPase inhibitor with Kd of 40 nM 50 nM 60 nM and 250 nM for Rac1 Rac1b Rac2 and Rac3 respectively