Learn More
Waters Corp AutoSpec EI Inner Ion Chamber
Supplier:  Waters Corp 700004772
Encompass_Preferred
Replacement EI Inner Chamber for an AutoSpec Premier. This is part of EI Source Assembly part number M805790CC1-S.
Catalog No. 50-466-616