Learn More
Thermo Scientific Ion Source Cartridge Kit
Catalog No. 501346489 Shop All Thermo Scientific Products
Encompass
Thermal switch for ion source assembly with bimetallic snap disc that opens electrical contact at predetermined temperature as safety mechanism to regulate temperature of ionization source in mass spectrometry systems, compatible with GC-MS.
This item is not returnable. View return policy
Catalog No. 50-134-6489 Supplier Thermo Scientific Supplier No. 1R1204044100
Thermal switch for ion source assembly with bimetallic snap disc that opens electrical contact at predetermined temperature as safety mechanism to regulate temperature of ionization source in mass spectrometry systems, compatible with GC-MS.