Learn More
Medchemexpress LLC HY-15573 5mg Medchemexpress, EI1 CAS:1418308-27-6 Purity:>98%
SDP
Catalog No. 502081741
Encompass_Preferred
Medchemexpress, HY-15573 5mg EI1 CAS:1418308-27-6 EI1 (KB-145943) is a potent and selective EZH2 inhibitor with IC50 of 15 nM and 13 nM for EZH2 (WT) and EZH2 (Y641F), respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1741 Supplier Medchemexpress LLC Supplier No. HY155735MG
Medchemexpress, HY-15573 5mg EI1 CAS:1418308-27-6 EI1 (KB-145943) is a potent and selective EZH2 inhibitor with IC50 of 15 nM and 13 nM for EZH2 (WT) and EZH2 (Y641F), respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.