Learn More
Abcam EI1, EZH2 inhibitor, 25MG
Catalog No. 502672013
Encompass_Preferred
MW 390.5 Da. Potent (IC₅₀ = 15 nM for wild-type enzyme; 13 nM for Y641F mutant enzyme) EZH2 inhibitor. Selective versus EZH1 (142-fold). Highly selective (>10,000-fold) against a panel of 10 other human methyltransferases. Inhibits cellular H3K27 methylation and activates Ezh2 target gene p16 expression in diffuse large B-cell lymphoma cells (DLBLC). Selectively inhibits growth and promotes cell cycle arrest and apoptosis of DLBLC cells with Y641 EZH2 mutations.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2013 Supplier Abcam Supplier No. AB26981725MG
MW 390.5 Da. Potent (IC₅₀ = 15 nM for wild-type enzyme; 13 nM for Y641F mutant enzyme) EZH2 inhibitor. Selective versus EZH1 (142-fold). Highly selective (>10,000-fold) against a panel of 10 other human methyltransferases. Inhibits cellular H3K27 methylation and activates Ezh2 target gene p16 expression in diffuse large B-cell lymphoma cells (DLBLC). Selectively inhibits growth and promotes cell cycle arrest and apoptosis of DLBLC cells with Y641 EZH2 mutations.
The product is subject to the following: Abcam Restricted Use Statement