Learn More
Aviva Systems Biology Corp EI24 Antibody - N-terminal region (ARP64952_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64952_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EI24. It was validated on Western Blot. . Alternate names: PIG8, TP53I8ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-288-398