Learn More
Bioworld EIA Buffer (10X), 100 mL
Catalog No. 50253028
Encompass_Preferred
EIA Buffer, 10X Solution A 1M Phosphate solution containing BSA, NaCl, EDTA, and Azide. Product contains 50mL of 10X solution concentrate and can be diluted with distilled water to create 500mL or 1X solution. Technical Information:pH 7.4Stability >1 year at 4° C
Catalog No. 50-253-028 Supplier Bioworld Supplier No. 220400302
EIA Buffer, 10X Solution A 1M Phosphate solution containing BSA, NaCl, EDTA, and Azide. Product contains 50mL of 10X solution concentrate and can be diluted with distilled water to create 500mL or 1X solution. Technical Information:pH 7.4Stability >1 year at 4° C