Learn More
Bioworld EIA Buffer (10X), 50 mL
Catalog No. 50253027
Encompass_Preferred
EIA Buffer, 10X Solution A 1M Phosphate solution containing BSA, NaCl, EDTA, and Azide. Product contains 50mL of 10X solution concentrate and can be diluted with distilled water to create 500mL or 1X solution. Technical Information:pH 7.4Stability >1 year at 4° C
Catalog No. 50-253-027 Supplier Bioworld Supplier No. 220400301
EIA Buffer, 10X Solution A 1M Phosphate solution containing BSA, NaCl, EDTA, and Azide. Product contains 50mL of 10X solution concentrate and can be diluted with distilled water to create 500mL or 1X solution. Technical Information:pH 7.4Stability >1 year at 4° C