Learn More
Eichrom Industries Extraction Chromatographic Resin, 50-100 μm particle size, 100 g bottle, Loose Resin, Bed Density 0.39 g/mL, Recommended Working Capacity 10 mg/mL U(VI)
SDP
Catalog No. NC3673116
Encompass
Extraction Chromatographic Resin, 50-100 μm particle size, 100 g bottle, Loose Resin, Bed Density 0.39 g/mL, Recommended Working Capacity 10 mg/mL U(VI)
Catalog No. NC3673116 Supplier Eichrom Industries Supplier No. UTB100S
Extraction Chromatographic Resin, 50-100 μm particle size, 100 g bottle, Loose Resin, Bed Density 0.39 g/mL, Recommended Working Capacity 10 mg/mL U(VI)