Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Eicosanedioic acid | 2424-92-2 | MFCD00059627 | 99.0% | 342.51 g/mol | C20H38O4 | 100 G
SDP

Catalog No. 5000204113
Encompass_Preferred

Eicosanedioic acid is a saturated C20 dicarboxylic acid used as a non-cleavable linker in antibody-drug conjugate (ADC) synthesis and as an alkyl chain-based linker for proteolysis-targeting chimera (PROTAC) construction. It is supplied with high purity for research and synthetic applications.

  • CAS number: 2424-92-2.
  • Chemical formula: C20H38O4.
  • Molecular weight: 342.51 g/mol.
  • Purity: 99.0%.
  • Available pack sizes: 5 g, 10 g, 25 g, 100 g, and bulk (quote for >100 g).
  • Common applications: ADC linker, PROTAC linker, synthetic intermediate.

Catalog No. 50-002-04113 Supplier Medchemexpress LLC Supplier No. HYW034595100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eicosanedioic acid is a saturated C20 dicarboxylic acid used as a non-cleavable linker in antibody-drug conjugate (ADC) synthesis and as an alkyl chain-based linker for proteolysis-targeting chimera (PROTAC) construction. It is supplied with high purity for research and synthetic applications.

  • CAS number: 2424-92-2.
  • Chemical formula: C20H38O4.
  • Molecular weight: 342.51 g/mol.
  • Purity: 99.0%.
  • Available pack sizes: 5 g, 10 g, 25 g, 100 g, and bulk (quote for >100 g).
  • Common applications: ADC linker, PROTAC linker, synthetic intermediate.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.