Learn More
American Radiolabeled Chemicals Inc EICOSAPENT 14C COA 10UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ARC 360010UC
Encompass_Preferred
Eicosapentaenoyl 5,8,11,14,17-1-14C Coenzyme A 10uCi in 0.1N HCl (pH 5.2), 0.1 mCi/ml, 50-60 mCi/mmol, M.W. 1069, Shipped in Dry Ice, Exclusive
Catalog No. 50-352-941