Learn More
Enzo Life Sciences Eicosapentaenoic acid (1g). CAS: 10417-94-4
SDP
Catalog No. 502010790
Encompass_Preferred
Beneficial effect in cardiovascular diseases and inflammation. Inhibits PGE2 formation from arachidonic acid in 3T3 fibroblats. Alternative name: Eicosa-5Z,8Z,11Z,14Z,17Z-pentaenoic acid (20:5, n-3), Icosapent. Purity: ≥98% (TLC). Solubility: Soluble in 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-201-0790 Supplier Enzo Life Sciences Supplier No. BMLFA0011000
Beneficial effect in cardiovascular diseases and inflammation. Inhibits PGE2 formation from arachidonic acid in 3T3 fibroblats. Alternative name: Eicosa-5Z,8Z,11Z,14Z,17Z-pentaenoic acid (20:5, n-3), Icosapent. Purity: ≥98% (TLC). Solubility: Soluble in 100% ethanol. Long Term Storage: -20°C.